Assay Details
Target Gene Details
Entrez Gene ID: | 101928085 |
Gene Name: | ERICH6 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:150703564-150720146 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERICH6-AS1 | NR_121674.1 | |||
| NR_121675.1 | ||||
| BP431953.1 | ||||
| DA374305.1 |

Genomic Map