Assay Details
Target Gene Details
Entrez Gene ID: | 84107 |
Gene Name: | Zic family member 4 |
Gene Aliases: |
- |
Location: |
Chr.3:147386048-147406893 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZIC4 | NM_001168378.1 | NP_001161850.1 | ||
| NM_001168379.1 | NP_001161851.1 | |||
| NM_001243256.1 | NP_001230185.1 | |||
| NM_032153.5 | NP_115529.2 | |||
| XM_017007314.1 | XP_016862803.1 | |||
| AK056262.1 | ||||
| AK293981.1 | ||||
| AK294831.1 | ||||
| BC029507.1 | AAH29507.1 | |||
| BC126267.1 | ||||
| BC136339.1 | ||||
| DA066220.1 | ||||
| DA218617.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv822285 | Chr.3:147381294 - 147428675 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115196329] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map