Assay Details
Target Gene Details
Entrez Gene ID: | 7095 |
Gene Name: | SEC62 homolog, preprotein translocation factor |
Gene Aliases: |
Dtrp1, HTP1, TLOC1, TP-1 |
Location: |
Chr.3:169966792-169998373 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEC62 | NM_003262.3 | NP_003253.1 | ||
| XM_011513114.2 | XP_011511416.1 | |||
| AK294504.1 | ||||
| BC003677.1 | ||||
| BC012035.1 | AAH12035.1 | |||
| BX475495.1 | ||||
| BX537668.1 | 1 | 5246 | ||
| BX648539.1 | ||||
| D87127.1 | BAA13254.1 | |||
| KC005990.1 | ||||
| U93239.1 | AAB51391.1 |
More Information
Set Membership: |
|

Genomic Map