Assay Details
Target Gene Details
Entrez Gene ID: | 22973 |
Gene Name: | laminin subunit beta 2 pseudogene 1 |
Gene Aliases: |
LAMB2L |
Location: |
Chr.3:49152859-49154401 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LAMB2P1 | NR_004405.1 | 1 | 795 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3394931 | Chr.3:49153115 - 49154713 on Build GRCh38 | Duplication |
|
| nsv834686 | Chr.3:49068614 - 49233123 on Build GRCh38 | Loss |
|

Genomic Map