Assay Details
Target Gene Details
Entrez Gene ID: | 51300 |
Gene Name: | translocase of inner mitochondrial membrane domain containing 1 |
Gene Aliases: |
C3orf1 |
Location: |
Chr.3:119498521-119524281 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TIMMDC1 | NM_016589.3 | NP_057673.2 | ||
| XM_017006556.1 | XP_016862045.1 | |||
| AF139077.1 | AAF62372.1 | |||
| AF210057.1 | AAG43510.1 | |||
| AK026250.1 | ||||
| AK225954.1 | ||||
| AL136622.1 | CAB66557.1 | |||
| AL390077.1 | CAB98201.1 | |||
| AL390090.1 | CAB98212.1 | |||
| AL390094.1 | CAB98251.1 | |||
| AY358633.1 | AAQ88996.1 | |||
| BC012341.1 | AAH12341.1 | |||
| BG705703.1 | ||||
| CR457158.1 | CAG33439.1 | |||
| CR533524.1 | CAG38555.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829695 | Chr.3:119460043 - 119649149 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map