Assay Details
Target Gene Details
Entrez Gene ID: | 9175 |
Gene Name: | mitogen-activated protein kinase kinase kinase 13 |
Gene Aliases: |
LZK, MEKK13, MLK |
Location: |
Chr.3:185282938-185489094 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAP3K13 | NM_001242314.1 | 15 | 4085 | NP_001229243.1 |
| NM_001242317.1 | 13 | 3349 | NP_001229246.1 | |
| NM_004721.4 | 14 | 4200 | NP_004712.1 | |
| XM_017007456.1 | 15 | 4096 | XP_016862945.1 |
Target Gene Details
Entrez Gene ID: | 90407 |
Gene Name: | transmembrane protein 41A |
Gene Aliases: |
2900010K02Rik |
Location: |
Chr.3:185482671-185499057 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM41A |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829818 | Chr.3:185411745 - 185567640 on Build GRCh38 | Loss |
|
| esv3893837 | Chr.3:185291669 - 185483550 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map