Assay Details
Target Gene Details
Entrez Gene ID: | 85403 |
Gene Name: | ELL associated factor 1 |
Gene Aliases: |
- |
Location: |
Chr.3:15427557-15442613 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EAF1 | NM_033083.6 | NP_149074.3 | ||
| XM_005265518.1 | XP_005265575.1 | |||
| XM_011534165.1 | XP_011532467.1 | |||
| XM_011534166.1 | XP_011532468.1 | |||
| AF272973.1 | AAK58687.1 | |||
| AK304697.1 | ||||
| AK312330.1 | ||||
| BC041329.2 | AAH41329.1 | |||
| BQ064584.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834622 | Chr.3:15397807 - 15587083 on Build GRCh38 | Gain |
|
| nsv834621 | Chr.3:15324275 - 15509456 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map