Assay Details
Target Gene Details
Entrez Gene ID: | 80012 |
Gene Name: | polyhomeotic homolog 3 |
Gene Aliases: |
EDR3, HPH3 |
Location: |
Chr.3:170087580-170181779 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv8728n54 | Chr.3:170056595 - 170215221 on Build GRCh38 | Loss |
|
| nsv1009884 | Chr.3:170001661 - 170189371 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map