Assay Details
Target Gene Details
Entrez Gene ID: | 64147 |
Gene Name: | kinesin family member 9 |
Gene Aliases: |
- |
Location: |
Chr.3:47228026-47282847 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIF9 | NM_001134878.1 | NP_001128350.1 | ||
| NM_022342.4 | NP_071737.1 | |||
| NM_182902.3 | NP_878905.2 | |||
| XM_005265388.4 | XP_005265445.1 | |||
| XM_005265389.4 | XP_005265446.1 | |||
| XM_006713291.3 | XP_006713354.1 | |||
| XM_011534003.2 | XP_011532305.1 | |||
| XM_011534004.2 | XP_011532306.1 | |||
| XM_011534005.2 | XP_011532307.1 | |||
| XM_011534006.2 | XP_011532308.1 | |||
| XM_011534007.2 | XP_011532309.1 | |||
| XM_017007029.1 | XP_016862518.1 | |||
| XM_017007030.1 | XP_016862519.1 | |||
| XM_017007031.1 | XP_016862520.1 | |||
| AF311212.1 | AAG33849.1 | |||
| AK292547.1 | ||||
| AK302070.1 | ||||
| AK314418.1 | ||||
| BC030657.2 | AAH30657.1 |
Target Gene Details
Entrez Gene ID: | 285352 |
Gene Name: | KIF9 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:47164370-47244116 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIF9-AS1 | NR_033373.1 | |||
| AK096853.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1007714 | Chr.3:47042897 - 47290080 on Build GRCh38 | Gain |
|
| nsv428417 | Chr.3:46638962 - 47778557 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map