Assay Details
Target Gene Details
Entrez Gene ID: | 80723 |
Gene Name: | solute carrier family 35 member G2 |
Gene Aliases: |
TMEM22 |
Location: |
Chr.3:136819019-136855892 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC35G2 | NM_001097599.1 | NP_001091068.1 | ||
| NM_001097600.1 | NP_001091069.1 | |||
| NM_025246.2 | NP_079522.2 | |||
| XM_017007290.1 | 1 | 219 | XP_016862779.1 | |
| AK096029.1 | 1 | 1813 | ||
| AK314079.1 | ||||
| AL136661.1 | CAB66596.1 | |||
| BC000111.1 | AAH00111.1 | |||
| BC022557.1 | AAH22557.1 | |||
| BI962644.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4027 | Chr.3:136807417 - 136831439 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map