Assay Details
Target Gene Details
Entrez Gene ID: | 165631 |
Gene Name: | poly(ADP-ribose) polymerase family member 15 |
Gene Aliases: |
ARTD7, BAL3, pART7 |
Location: |
Chr.3:122575914-122639047 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PARP15 | NM_001113523.2 | NP_001106995.1 | ||
| NM_001308320.1 | NP_001295249.1 | |||
| XM_005247159.3 | XP_005247216.1 | |||
| XM_005247160.3 | XP_005247217.1 | |||
| XM_011512475.2 | XP_011510777.1 | |||
| XM_011512476.2 | XP_011510778.1 | |||
| XM_011512477.2 | XP_011510779.1 | |||
| XM_011512478.2 | XP_011510780.1 | |||
| XM_011512479.2 | XP_011510781.1 | |||
| XM_011512480.2 | XP_011510782.1 | |||
| XM_017005791.1 | XP_016861280.1 | |||
| XM_017005792.1 | 1 | 582 | XP_016861281.1 | |
| AK097515.1 | ||||
| AK309863.1 | ||||
| BX647656.1 | ||||
| DB158535.1 | ||||
| DQ063586.1 | AAY64451.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829705 | Chr.3:122595197 - 122741131 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map