Assay Details
Target Gene Details
Entrez Gene ID: | 9197 |
Gene Name: | solute carrier family 33 member 1 |
Gene Aliases: |
ACATN, AT-1, AT1, CCHLND, SPG42 |
Location: |
Chr.3:155826511-155854459 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC33A1 | NM_001190992.1 | 7 | 2294 | NP_001177921.1 |
| NM_004733.3 | 6 | 3656 | NP_004724.1 | |
| XM_011513311.2 | 4 | 3323 | XP_011511613.1 | |
| XM_017007462.1 | 4 | 3320 | XP_016862951.1 | |
| XM_017007463.1 | 6 | 3739 | XP_016862952.1 | |
| XM_017007464.1 | 5 | 3425 | XP_016862953.1 | |
| BC014416.1 | 7 | 2241 | AAH14416.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3598283 | Chr.3:155759067 - 155869415 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map