Assay Details
Target Gene Details
Entrez Gene ID: | 7104 |
Gene Name: | transmembrane 4 L six family member 4 |
Gene Aliases: |
ILTMP, il-TMP |
Location: |
Chr.3:149474581-149503394 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TM4SF4 | NM_004617.3 | NP_004608.1 | ||
| AK055577.1 | ||||
| AK315464.1 | ||||
| BC001386.2 | AAH01386.1 | |||
| BT007430.1 | AAP36098.1 | |||
| U31449.1 | AAA74745.1 |
More Information
Set Membership: |
|

Genomic Map