Assay Details
Target Gene Details
Entrez Gene ID: | 3059 |
Gene Name: | hematopoietic cell-specific Lyn substrate 1 |
Gene Aliases: |
CTTNL, HS1, lckBP1, p75 |
Location: |
Chr.3:121631398-121660948 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HCLS1 | NM_001292041.1 | NP_001278970.1 | ||
| NM_005335.5 | NP_005326.2 | |||
| AK225633.1 | ||||
| AK298663.1 | ||||
| AK298696.1 | ||||
| AK300301.1 | ||||
| AK312750.1 | ||||
| BC016758.2 | AAH16758.1 | |||
| BT006824.1 | AAP35470.1 | |||
| CR456794.1 | CAG33075.1 | |||
| DC407066.1 | ||||
| DC427610.1 | ||||
| X16663.1 | CAA34651.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1007522 | Chr.3:121481716 - 121655204 on Build GRCh38 | Gain |
|
| dgv8524n54 | Chr.3:121252053 - 121838686 on Build GRCh38 | Gain |
|
| esv3575599 | Chr.3:121571453 - 121671424 on Build GRCh38 | Gain |
|
| nsv998740 | Chr.3:121570652 - 121671424 on Build GRCh38 | Gain |
|
| esv3575598 | Chr.3:121406926 - 121715587 on Build GRCh38 | Gain |
|
| nsv999335 | Chr.3:121381436 - 121689919 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map