Assay Details
Target Gene Details
Entrez Gene ID: | 285381 |
Gene Name: | diphthamide biosynthesis 3 |
Gene Aliases: |
DELGIP, DELGIP1, DESR1, DPH3A, KTI11, ZCSL2 |
Location: |
Chr.3:16257061-16264989 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DPH3 | NM_001047434.2 | 2 | 2717 | NP_001040899.1 |
| NM_206831.2 | 3 | 2792 | NP_996662.1 | |
| AK022970.1 | 2 | 2697 | ||
| AK023203.1 | 1 | 1717 | ||
| AK092040.1 | 3 | 2745 |
More Information
Set Membership: |
|

Genomic Map