Assay Details
Target Gene Details
Entrez Gene ID: | 80012 |
Gene Name: | polyhomeotic homolog 3 |
Gene Aliases: |
EDR3, HPH3 |
Location: |
Chr.3:170087580-170181779 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHC3 | NM_001308116.1 | 15 | 11973 | NP_001295045.1 |
| NM_024947.3 | 15 | 11958 | NP_079223.3 | |
| XM_005247789.2 | 16 | 12036 | XP_005247846.1 | |
| XM_006713753.1 | 16 | 12005 | XP_006713816.1 | |
| XM_006713754.3 | 16 | 12127 | XP_006713817.1 | |
| XM_006713756.2 | 14 | 11778 | XP_006713819.1 | |
| XM_011513186.2 | 15 | 11937 | XP_011511488.1 | |
| XM_011513187.2 | 14 | 11889 | XP_011511489.1 | |
| XM_011513188.2 | 15 | 11877 | XP_011511490.1 | |
| XM_017007235.1 | 16 | 12024 | XP_016862724.1 | |
| XM_017007236.1 | 16 | 12012 | XP_016862725.1 | |
| XM_017007237.1 | 14 | 11829 | XP_016862726.1 | |
| XM_017007238.1 | 13 | 11730 | XP_016862727.1 | |
| XM_017007239.1 | 13 | 11577 | XP_016862728.1 | |
| BX647868.1 | 8 | 2552 | CAI46090.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv8728n54 | Chr.3:170056595 - 170215221 on Build GRCh38 | Loss |
|
| nsv1009884 | Chr.3:170001661 - 170189371 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map