Assay Details
Target Gene Details
Entrez Gene ID: | 7879 |
Gene Name: | RAB7A, member RAS oncogene family |
Gene Aliases: |
PRO2706, RAB7 |
Location: |
Chr.3:128726136-128814798 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB7A | NM_004637.5 | NP_004628.4 | ||
| AK094449.1 | ||||
| AK290721.1 | ||||
| AK298344.1 | ||||
| AK308934.1 | ||||
| AU122058.1 | ||||
| BC008721.2 | AAH08721.2 |
More Information
Set Membership: |
|

Genomic Map