Assay Details
Target Gene Details
Entrez Gene ID: | 100128640 |
Gene Name: | ACVR2B antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:38451027-38454820 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACVR2B-AS1 | NR_028389.1 | 2 | 1290 | |
| AK025619.1 | 2 | 1290 |
Target Gene Details
Entrez Gene ID: | 9942 |
Gene Name: | xylulokinase |
Gene Aliases: |
- |
Location: |
Chr.3:38346760-38453041 on Build GRCh38 |
Assay Gene Location: | Within Intron 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| XYLB | XM_011534327.1 | XP_011532629.1 | ||
| XM_011534329.1 | XP_011532631.1 |
More Information
Set Membership: |
|

Genomic Map