Assay Details
Target Gene Details
Entrez Gene ID: | 8833 |
Gene Name: | guanine monophosphate synthase |
Gene Aliases: |
- |
Location: |
Chr.3:155869430-155943679 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GMPS | NM_003875.2 | NP_003866.1 | ||
| XM_011513263.2 | XP_011511565.1 | |||
| XM_017007422.1 | XP_016862911.1 | |||
| AK223399.1 | BAD97119.1 | |||
| AK291504.1 | ||||
| AK300787.1 | ||||
| AK302148.1 | ||||
| AK315832.1 | ||||
| BC012178.1 | AAH12178.1 | |||
| BG721087.1 | ||||
| U10860.1 | AAA60331.1 |
More Information
Set Membership: |
|

Genomic Map