Assay Details
Target Gene Details
Entrez Gene ID: | 686 |
Gene Name: | biotinidase |
Gene Aliases: |
- |
Location: |
Chr.3:15601352-15653709 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BTD | NM_001281726.1 | NP_001268655.1 | ||
| XM_006713314.3 | XP_006713377.1 | |||
| XM_011534041.2 | XP_011532343.1 | |||
| XM_017007088.1 | XP_016862577.1 | |||
| AK294301.1 | ||||
| AK297033.1 | ||||
| AK301838.1 | ||||
| AK313252.1 | ||||
| BC012099.1 | AAH12099.1 | |||
| BE814688.1 | ||||
| BE814713.1 | ||||
| BM839350.1 | ||||
| BM919775.1 | ||||
| BP326502.1 | ||||
| U03274.1 | AAC04318.1 |
More Information
Set Membership: |
|

Genomic Map