Assay Details
Target Gene Details
Entrez Gene ID: | 51088 |
Gene Name: | kelch like family member 5 |
Gene Aliases: |
- |
Location: |
Chr.4:39044796-39143101 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLHL5 | NM_001007075.2 | NP_001007076.1 | ||
| NM_001171654.1 | NP_001165125.1 | |||
| NM_015990.4 | NP_057074.3 | |||
| NM_199039.3 | NP_950240.2 | |||
| XM_005262655.4 | XP_005262712.1 | |||
| XM_011513699.2 | XP_011512001.1 | |||
| XM_011513700.2 | XP_011512002.1 | |||
| XM_011513701.2 | XP_011512003.1 | |||
| XM_017008273.1 | XP_016863762.1 | |||
| XM_017008274.1 | XP_016863763.1 | |||
| XM_017008275.1 | XP_016863764.1 | |||
| XM_017008276.1 | XP_016863765.1 | |||
| AB208820.1 | BAD92057.1 | |||
| AF272976.1 | AAL08584.1 | |||
| AK289785.1 | ||||
| AY172948.1 | AAO39710.1 | |||
| BC053860.1 | AAH53860.1 | |||
| DB451501.1 | ||||
| DR006899.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs114228407] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map