Assay Details
Target Gene Details
Entrez Gene ID: | 80167 |
Gene Name: | abhydrolase domain containing 18 |
Gene Aliases: |
C4orf29 |
Location: |
Chr.4:127965256-128039711 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABHD18 | NM_001319305.1 | NP_001306234.1 | ||
| NM_001319306.1 | NP_001306235.1 | |||
| NM_001319307.1 | NP_001306236.1 | |||
| XM_005263238.4 | XP_005263295.1 | |||
| XM_006714318.3 | XP_006714381.1 | |||
| XM_006714320.3 | XP_006714383.1 | |||
| XM_011532272.2 | XP_011530574.1 | |||
| XM_011532273.2 | XP_011530575.1 | |||
| XM_011532274.2 | XP_011530576.1 | |||
| XM_011532276.1 | XP_011530578.1 | |||
| XM_011532278.2 | XP_011530580.1 | |||
| XM_017008631.1 | XP_016864120.1 | |||
| AK024759.1 | BAB14990.1 | |||
| BC034253.1 | ||||
| BC128142.1 | ||||
| BC128143.1 | ||||
| BX500590.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5385n100 | Chr.4:127882004 - 128173863 on Build GRCh38 | Gain |
|
| nsv595388 | Chr.4:127932758 - 128163676 on Build GRCh38 | Gain |
|
| nsv1019193 | Chr.4:127889372 - 128028165 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs138335795] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map