Assay Details
Target Gene Details
Entrez Gene ID: | 55013 |
Gene Name: | mitochondrial calcium uniporter dominant negative beta subunit |
Gene Aliases: |
CCDC109B |
Location: |
Chr.4:109560199-109688719 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCUB | NM_017918.4 | NP_060388.2 | ||
| XM_006714246.3 | XP_006714309.1 | |||
| AK095936.1 | ||||
| AK291098.1 | ||||
| AK311030.1 | ||||
| BC002633.2 | AAH02633.1 | |||
| CN427680.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv595129 | Chr.4:109000908 - 109672304 on Build GRCh38 | Gain |
|
| esv3894003 | Chr.4:109617846 - 109642294 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map