Assay Details
Target Gene Details
Entrez Gene ID: | 54532 |
Gene Name: | ubiquitin specific peptidase 53 |
Gene Aliases: |
- |
Location: |
Chr.4:119212583-119295518 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP53 | NM_019050.2 | NP_061923.2 | ||
| XM_005263073.3 | XP_005263130.1 | |||
| XM_005263075.2 | XP_005263132.1 | |||
| XM_005263076.2 | XP_005263133.1 | |||
| XM_005263077.3 | XP_005263134.1 | |||
| XM_006714240.1 | XP_006714303.1 | |||
| XM_011532037.2 | XP_011530339.1 | |||
| XM_011532038.2 | XP_011530340.1 | |||
| XM_017008312.1 | XP_016863801.1 | |||
| XM_017008313.1 | XP_016863802.1 | |||
| XM_017008314.1 | XP_016863803.1 | |||
| XM_017008315.1 | XP_016863804.1 | |||
| XM_017008316.1 | XP_016863805.1 | |||
| XM_017008317.1 | XP_016863806.1 | |||
| XM_017008318.1 | XP_016863807.1 | |||
| CR749490.1 | CAH18315.1 | |||
| DA859085.1 | ||||
| DA886370.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2763386 | Chr.4:119222706 - 119265813 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map