Assay Details
Target Gene Details
Entrez Gene ID: | 8819 |
Gene Name: | Sin3A associated protein 30 |
Gene Aliases: |
- |
Location: |
Chr.4:173370942-173377532 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SAP30 | NM_003864.3 | NP_003855.1 | ||
| AF055993.1 | AAC33316.1 | |||
| BC016757.1 | AAH16757.1 |
More Information
Set Membership: |
|

Genomic Map