Assay Details
Target Gene Details
Entrez Gene ID: | 3815 |
Gene Name: | KIT proto-oncogene receptor tyrosine kinase |
Gene Aliases: |
C-Kit, CD117, PBT, SCFR |
Location: |
Chr.4:54657928-54740715 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIT | NM_000222.2 | NP_000213.1 | ||
| NM_001093772.1 | NP_001087241.1 | |||
| XM_005265740.1 | XP_005265797.1 | |||
| XM_005265741.1 | XP_005265798.1 | |||
| XM_005265742.2 | XP_005265799.1 | |||
| XM_017008178.1 | XP_016863667.1 | |||
| XM_017008179.1 | XP_016863668.1 | |||
| XM_017008180.1 | XP_016863669.1 | |||
| AK304031.1 | ||||
| BC071593.1 | AAH71593.1 | |||
| DC376760.1 | ||||
| EU826594.1 | ||||
| GU983671.1 | ||||
| HM015525.1 | ||||
| HM015526.1 | ||||
| KT326922.1 | ||||
| X06182.1 | CAA29548.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526344 | Chr.4:54656321 - 55364952 on Build GRCh38 | Gain |
|
| nsv1009516 | Chr.4:54682283 - 54694673 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73234236] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map