Assay Details
Target Gene Details
Entrez Gene ID: | 3818 |
Gene Name: | kallikrein B1 |
Gene Aliases: |
KLK3, PKK, PKKD, PPK |
Location: |
Chr.4:186215714-186258477 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLKB1 | XM_017008181.1 | 1 | 256 | XP_016863670.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1028146 | Chr.4:185582745 - 186417181 on Build GRCh38 | Gain |
|
| nsv4644 | Chr.4:186185600 - 186219291 on Build GRCh38 | Insertion |
|
| nsv830172 | Chr.4:186091611 - 186247270 on Build GRCh38 | Loss |
|
| nsv461864 | Chr.4:186085652 - 186335850 on Build GRCh38 | Gain |
|
| nsv596401 | Chr.4:186151229 - 186445495 on Build GRCh38 | Loss |
|
| esv3603525 | Chr.4:186210472 - 186225457 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map