Assay Details
Target Gene Details
Entrez Gene ID: | 7222 |
Gene Name: | transient receptor potential cation channel subfamily C member 3 |
Gene Aliases: |
SCA41, TRP3 |
Location: |
Chr.4:121879028-121952060 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRPC3 | NM_001130698.1 | NP_001124170.1 | ||
| NM_003305.2 | NP_003296.1 | |||
| XM_011532217.2 | XP_011530519.1 | |||
| XM_011532218.2 | XP_011530520.1 | |||
| XM_017008578.1 | XP_016864067.1 | |||
| XM_017008579.1 | XP_016864068.1 | |||
| AB255423.1 | ||||
| AB255424.1 | ||||
| AY865574.1 | AAW62292.1 | |||
| BC093682.1 | AAH93682.1 | |||
| BC093684.1 | AAH93684.1 | |||
| KC207570.1 | ||||
| Y13758.1 | CAA74083.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv997539 | Chr.4:121622949 - 122353111 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map