Assay Details
Target Gene Details
Entrez Gene ID: | 339965 |
Gene Name: | coiled-coil domain containing 158 |
Gene Aliases: |
- |
Location: |
Chr.4:76313039-76421920 on Build GRCh38 |
Assay Gene Location: | Within Intron 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC158 | NM_001042784.1 | NP_001036249.1 | ||
| XM_005262974.4 | XP_005263031.1 | |||
| XM_006714204.3 | XP_006714267.1 | |||
| XM_011531908.2 | XP_011530210.1 | |||
| XM_011531909.2 | XP_011530211.1 | |||
| XM_011531910.2 | XP_011530212.1 | |||
| XM_011531911.1 | XP_011530213.1 | |||
| XM_011531912.2 | XP_011530214.1 | |||
| XM_011531913.1 | XP_011530215.1 | |||
| XM_011531914.2 | XP_011530216.1 | |||
| XM_011531915.2 | XP_011530217.1 | |||
| XM_011531916.2 | XP_011530218.1 | |||
| XM_011531917.1 | XP_011530219.1 | |||
| XM_017008160.1 | XP_016863649.1 | |||
| XM_017008161.1 | XP_016863650.1 | |||
| AK098600.1 | BAC05347.1 | |||
| AK098636.1 | BAC05359.1 | |||
| AL832445.1 | ||||
| BC086869.1 | AAH86869.1 |

Genomic Map