Assay Details
Target Gene Details
Entrez Gene ID: | 2743 |
Gene Name: | glycine receptor beta |
Gene Aliases: |
HKPX2 |
Location: |
Chr.4:157076125-157172090 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLRB | NM_000824.4 | NP_000815.1 | ||
| NM_001166060.1 | NP_001159532.1 | |||
| NM_001166061.1 | NP_001159533.1 | |||
| XM_017008035.1 | XP_016863524.1 | |||
| AF094754.1 | AAC71033.1 | |||
| AF094755.1 | AAC71034.1 | |||
| AK290617.1 | ||||
| BC032635.1 | AAH32635.1 | |||
| DA168145.1 | ||||
| DA528659.1 | ||||
| DB479295.1 | ||||
| U33267.1 | AAB37750.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1027454 | Chr.4:157056878 - 157141746 on Build GRCh38 | Gain |
|
| nsv1018387 | Chr.4:157038779 - 157843038 on Build GRCh38 | Gain |
|
| nsv595803 | Chr.4:157028236 - 157171909 on Build GRCh38 | Gain |
|
| esv3602801 | Chr.4:156958946 - 157162701 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs80301105] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map