Assay Details
Target Gene Details
Entrez Gene ID: | 153642 |
Gene Name: | arylsulfatase family member K |
Gene Aliases: |
TSULF |
Location: |
Chr.5:95555108-95605102 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARSK | NM_198150.2 | NP_937793.1 | ||
| XM_005271904.3 | XP_005271961.1 | |||
| AK303855.1 | ||||
| AK316253.1 | ||||
| AL832711.1 | CAD38634.1 | |||
| AY358596.1 | AAQ88959.1 | |||
| AY875939.1 | AAW66667.1 | |||
| BC036047.1 | AAH36047.1 | |||
| BC050392.1 | AAH50392.1 | |||
| BC130329.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv598955 | Chr.5:95567749 - 95800315 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map