Assay Details
Target Gene Details
Entrez Gene ID: | 133522 |
Gene Name: | PPARG coactivator 1 beta |
Gene Aliases: |
ERRL1, PERC, PGC-1(beta), PGC1B |
Location: |
Chr.5:149730252-149857861 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPARGC1B | NM_001172698.1 | NP_001166169.1 | ||
| NM_133263.3 | NP_573570.3 | |||
| XM_011537553.2 | XP_011535855.1 | |||
| XM_011537555.2 | XP_011535857.1 | |||
| XM_011537557.1 | XP_011535859.1 | |||
| AF468496.1 | AAL78633.1 | |||
| AF468497.1 | AAL78634.1 | |||
| AK095391.1 | BAC04541.1 | |||
| AY188947.1 | AAO40022.1 | |||
| AY188948.1 | AAO40023.1 | |||
| AY188949.1 | AAO40024.1 | |||
| AY188950.1 | AAO40025.1 | |||
| BC132971.1 | ||||
| BC132973.1 | ||||
| BC144251.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5060 | Chr.5:149693490 - 149739293 on Build GRCh38 | Deletion |
|
| nsv1019813 | Chr.5:149673743 - 149781874 on Build GRCh38 | Gain |
|
| nsv1027896 | Chr.5:149697049 - 149739278 on Build GRCh38 | Gain |
|
| nsv1073923 | Chr.5:149731236 - 149732137 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map