Assay Details
Target Gene Details
Entrez Gene ID: | 995 |
Gene Name: | cell division cycle 25C |
Gene Aliases: |
CDC25, PPP1R60 |
Location: |
Chr.5:138285265-138338355 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1033604 | Chr.5:138291351 - 138404936 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map