Assay Details
Target Gene Details
Entrez Gene ID: | 55278 |
Gene Name: | glutaminyl-tRNA synthase (glutamine-hydrolyzing)-like 1 |
Gene Aliases: |
GatA |
Location: |
Chr.6:106629566-106668417 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| QRSL1 | NM_018292.4 | NP_060762.3 | ||
| XM_011535924.2 | XP_011534226.1 | |||
| AK001851.1 | BAA91941.1 | |||
| AK023509.1 | BAB14592.1 | |||
| AK225262.1 | ||||
| AK225443.1 | ||||
| AK314694.1 | ||||
| AL136679.1 | CAB66614.1 | |||
| BC006084.1 | AAH06084.1 | |||
| BC014389.2 | AAH14389.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3610281 | Chr.6:106659033 - 106662913 on Build GRCh38 | Loss |
|
| nsv516902 | Chr.6:106656499 - 106667519 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map