Assay Details
Target Gene Details
Entrez Gene ID: | 134829 |
Gene Name: | clavesin 2 |
Gene Aliases: |
C6orf212, C6orf213, RLBP1L2, bA160A10.4 |
Location: |
Chr.6:122993693-123081282 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLVS2 | NM_001010852.3 | NP_001010852.2 | ||
| XM_017010266.1 | XP_016865755.1 | |||
| AK095527.1 | ||||
| AK295158.1 | ||||
| BC101376.1 | AAI01377.1 | |||
| BC101377.2 |
More Information
Set Membership: |
|

Genomic Map