Assay Details
Target Gene Details
Entrez Gene ID: | 55633 |
Gene Name: | TBC1 domain family member 22B |
Gene Aliases: |
C6orf197 |
Location: |
Chr.6:37257704-37332970 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBC1D22B | NM_017772.3 | NP_060242.2 | ||
| NR_130108.1 | ||||
| XM_011514738.2 | XP_011513040.1 | |||
| XM_011514739.1 | XP_011513041.1 | |||
| AB449909.1 | ||||
| AK096340.1 | ||||
| AK292893.1 | ||||
| AY313781.1 | AAQ72548.1 | |||
| BC109026.1 | AAI09027.1 | |||
| BC109027.1 | AAI09028.1 | |||
| BX648317.1 | ||||
| DA726945.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1015689 | Chr.6:36550916 - 39052434 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map