Assay Details
Target Gene Details
Entrez Gene ID: | 93663 |
Gene Name: | Rho GTPase activating protein 18 |
Gene Aliases: |
MacGAP, SENEX, bA307O14.2 |
Location: |
Chr.6:129576145-129710225 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP18 | XM_005267212.2 | 15 | 4004 | XP_005267269.2 |
| XM_005267213.1 | 14 | 3803 | XP_005267270.1 | |
| XM_017011494.1 | 14 | 3879 | XP_016866983.1 | |
| AK002195.1 | 2 | 1966 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv604663 | Chr.6:129482340 - 130671010 on Build GRCh38 | Gain |
|
| nsv524782 | Chr.6:129575356 - 129724532 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114073692] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map