Assay Details
Target Gene Details
Entrez Gene ID: | 54901 |
Gene Name: | CDK5 regulatory subunit associated protein 1 like 1 |
Gene Aliases: |
- |
Location: |
Chr.6:20534457-21232404 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDKAL1 | NM_017774.3 | NP_060244.2 | ||
| XM_011514718.1 | XP_011513020.1 | |||
| XM_011514719.2 | XP_011513021.1 | |||
| XM_017010986.1 | XP_016866475.1 | |||
| XM_017010987.1 | XP_016866476.1 | |||
| AK000349.1 | BAA91102.1 | |||
| AK128546.1 | BAC87494.1 | |||
| AK291735.1 | ||||
| AK309457.1 | ||||
| AK310219.1 | ||||
| BC064145.1 | AAH64145.1 | |||
| BC121020.1 | ||||
| BC121021.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1024963 | Chr.6:20474387 - 20862380 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map