Assay Details
Target Gene Details
Entrez Gene ID: | 101927703 |
Gene Name: | FOXC1 upstream transcript (non-protein coding) |
Gene Aliases: |
LINC01379, TCONS_00011636 |
Location: |
Chr.6:1605531-1607358 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOXCUT | NR_125804.1 | 2 | 897 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv992n67 | Chr.6:1589856 - 1629286 on Build GRCh38 | Gain |
|
| nsv1016859 | Chr.6:1459785 - 1645585 on Build GRCh38 | Gain |
|
| nsv823397 | Chr.6:1603632 - 1622029 on Build GRCh38 | Gain |
|

Genomic Map