Assay Details
Target Gene Details
Entrez Gene ID: | 23294 |
Gene Name: | ankyrin repeat and sterile alpha motif domain containing 1A |
Gene Aliases: |
ANKS1 |
Location: |
Chr.6:34889261-35097994 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKS1A | NM_015245.2 | NP_056060.2 | ||
| XM_005248964.3 | XP_005249021.1 | |||
| XM_006715036.2 | XP_006715099.1 | |||
| XM_011514431.2 | XP_011512733.1 | |||
| XM_011514432.2 | XP_011512734.1 | |||
| XM_011514433.1 | XP_011512735.1 | |||
| XM_011514434.2 | XP_011512736.1 | |||
| XM_011514435.2 | XP_011512737.1 | |||
| AK298987.1 | ||||
| BC022396.1 | ||||
| BC031934.1 | AAH31934.1 | |||
| BC132832.1 | ||||
| D86982.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508402 | Chr.6:34873184 - 34983511 on Build GRCh38 | Deletion |
|
| nsv1026121 | Chr.6:34819110 - 34993583 on Build GRCh38 | Gain |
|
| nsv462905 | Chr.6:34760476 - 34918659 on Build GRCh38 | Gain |
|
| dgv5955n100 | Chr.6:34755946 - 34919539 on Build GRCh38 | Gain |
|
| esv2763967 | Chr.6:34598478 - 34957686 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs59493795] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map