Assay Details
Target Gene Details
Entrez Gene ID: | 2037 |
Gene Name: | erythrocyte membrane protein band 4.1 like 2 |
Gene Aliases: |
4.1-G, 4.1G |
Location: |
Chr.6:130829111-131063322 on Build GRCh38 |
Assay Gene Location: | Within Intron 24 |
More Information
Additional Information:
For this assay, SNP(s) [rs111721346] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map