Assay Details
Target Gene Details
Entrez Gene ID: | 5961 |
Gene Name: | peripherin 2 |
Gene Aliases: |
AOFMD, AVMD, CACD2, DS, MDBS1, PRPH, RDS, RP7, TSPAN22, rd2 |
Location: |
Chr.6:42694509-42723024 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRPH2 | NM_000322.4 | NP_000313.2 | ||
| BC074720.2 | AAH74720.1 | |||
| M62958.1 | ||||
| M73531.1 | AAA60260.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830650 | Chr.6:42690493 - 42873400 on Build GRCh38 | Loss |
|
| nsv1017398 | Chr.6:42659705 - 43400998 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75499937] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map