Assay Details
Target Gene Details
Entrez Gene ID: | 222584 |
Gene Name: | family with sequence similarity 83 member B |
Gene Aliases: |
C6orf143 |
Location: |
Chr.6:54846323-54945099 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM83B | NM_001010872.2 | NP_001010872.1 | ||
| XM_006715022.3 | XP_006715085.1 | |||
| XM_017010478.1 | XP_016865967.1 | |||
| AK055204.1 | BAB70873.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1161381 | Chr.6:54830076 - 54868713 on Build GRCh38 | Duplication |
|
| esv2763553 | Chr.6:54823181 - 54955689 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs550767462] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map