Assay Details
Target Gene Details
Entrez Gene ID: | 57038 |
Gene Name: | arginyl-tRNA synthetase 2, mitochondrial |
Gene Aliases: |
ArgRS, DALRD2, PCH6, PRO1992, RARSL |
Location: |
Chr.6:87513744-87590032 on Build GRCh38 |
Assay Gene Location: | Within Intron 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RARS2 | NM_001318785.1 | NP_001305714.1 | ||
| NM_020320.4 | NP_064716.2 | |||
| NR_134857.1 | ||||
| XM_011535948.2 | XP_011534250.1 | |||
| XM_017011072.1 | XP_016866561.1 | |||
| XM_017011073.1 | XP_016866562.1 | |||
| XM_017011074.1 | XP_016866563.1 | |||
| XM_017011075.1 | XP_016866564.1 | |||
| XM_017011076.1 | XP_016866565.1 | |||
| XM_017011077.1 | XP_016866566.1 | |||
| XM_017011078.1 | XP_016866567.1 | |||
| XM_017011079.1 | XP_016866568.1 | |||
| XM_017011080.1 | XP_016866569.1 | |||
| XM_017011081.1 | XP_016866570.1 | |||
| AK023550.1 | BAB14608.1 | |||
| AK093934.1 | ||||
| AK315669.1 | ||||
| BC010420.1 | AAH10420.1 | |||
| CA314872.1 | ||||
| CA415020.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv6085n100 | Chr.6:87443361 - 87700332 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map