Assay Details
Target Gene Details
Entrez Gene ID: | 10492 |
Gene Name: | synaptotagmin binding cytoplasmic RNA interacting protein |
Gene Aliases: |
GRY-RBP, GRYRBP, HNRNPQ, HNRPQ1, NSAP1, PP68, hnRNP-Q |
Location: |
Chr.6:85607784-85643862 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5383 | Chr.6:85593505 - 85638305 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map