Assay Details
Target Gene Details
Entrez Gene ID: | 729178 |
Gene Name: | STXBP5 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.6:146841388-147204614 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STXBP5-AS1 | NR_034115.1 | 11 | 2034 | |
| AK125216.1 | 6 | 1493 | ||
| BQ269903.1 | 1 | 331 | ||
| DB318281.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830838 | Chr.6:146769609 - 146953533 on Build GRCh38 | Gain |
|

Genomic Map