Assay Details
Target Gene Details
Entrez Gene ID: | 285761 |
Gene Name: | discoidin, CUB and LCCL domain containing 1 |
Gene Aliases: |
dJ94G16.1 |
Location: |
Chr.6:117480346-117569858 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCBLD1 | NM_173674.2 | NP_775945.1 | ||
| XM_005266941.1 | XP_005266998.1 | |||
| XM_006715463.1 | XP_006715526.1 | |||
| XM_011535771.1 | XP_011534073.1 | |||
| XM_011535773.2 | XP_011534075.1 | |||
| XM_011535774.2 | XP_011534076.1 | |||
| XM_017010775.1 | XP_016866264.1 | |||
| AK095973.1 | BAC04663.1 | |||
| BC035671.1 | AAH35671.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs75911582] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map