Assay Details
Target Gene Details
Entrez Gene ID: | 2037 |
Gene Name: | erythrocyte membrane protein band 4.1 like 2 |
Gene Aliases: |
4.1-G, 4.1G |
Location: |
Chr.6:130829111-131063322 on Build GRCh38 |
Assay Gene Location: | Within Intron 25 |
More Information
Set Membership: |
|

Genomic Map