Assay Details
Target Gene Details
Entrez Gene ID: | 22881 |
Gene Name: | ankyrin repeat domain 6 |
Gene Aliases: |
- |
Location: |
Chr.6:89433178-89633834 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD6 | NM_001242809.1 | NP_001229738.1 | ||
| NM_001242813.1 | NP_001229742.1 | |||
| NM_014942.4 | NP_055757.3 | |||
| XM_011535598.2 | XP_011533900.1 | |||
| XM_011535599.2 | XP_011533901.1 | |||
| XM_011535600.2 | XP_011533902.1 | |||
| XM_017010495.1 | XP_016865984.1 | |||
| XM_017010498.1 | XP_016865987.1 | |||
| XM_017010501.1 | XP_016865990.1 | |||
| XM_017010505.1 | XP_016865994.1 | |||
| XM_017010510.1 | XP_016865999.1 | |||
| XM_017010512.1 | XP_016866001.1 | |||
| XM_017010514.1 | XP_016866003.1 | |||
| AB023174.1 | ||||
| AK307792.1 | ||||
| BC042173.1 | AAH42173.1 | |||
| DN999929.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3890901 | Chr.6:89204468 - 90033564 on Build GRCh38 | Gain |
|
| esv3609904 | Chr.6:89453348 - 89606022 on Build GRCh38 | Gain |
|
| dgv6089n100 | Chr.6:89446386 - 89678417 on Build GRCh38 | Gain |
|
| esv3890902 | Chr.6:89473670 - 89492658 on Build GRCh38 | Loss |
|
| nsv1027007 | Chr.6:89099218 - 89628146 on Build GRCh38 | Gain |
|
| esv3609902 | Chr.6:89444117 - 89509551 on Build GRCh38 | Gain |
|
| esv3609903 | Chr.6:89446569 - 89685111 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map