Assay Details
Target Gene Details
Entrez Gene ID: | 84624 |
Gene Name: | fibronectin type III domain containing 1 |
Gene Aliases: |
AGS8, FNDC2, MEL4B3, bA243O10.1, dJ322A24.1 |
Location: |
Chr.6:159169397-159272108 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FNDC1 | NM_032532.2 | NP_115921.2 | ||
| XM_011536190.2 | XP_011534492.1 | |||
| XM_011536191.2 | XP_011534493.1 | |||
| AB058769.2 | BAB47495.2 | |||
| BC146783.1 | ||||
| BC150607.1 | ||||
| DQ009660.1 | AAY26234.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3611412 | Chr.6:159202231 - 159205661 on Build GRCh38 | Loss |
|
| esv2763604 | Chr.6:159088341 - 159521923 on Build GRCh38 | Gain |
|
| nsv1019191 | Chr.6:159134068 - 159622215 on Build GRCh38 | Gain |
|
| nsv830850 | Chr.6:159163229 - 159357812 on Build GRCh38 | Gain |
|

Genomic Map